4,4'-Methylenedi-o-toluidine ≥97.0% (by titrimetric analysis)

Leverandør: TCI
Danger

D2046-25G
TCIAD2046-25GEA 3650 NOK
TCIAD2046-25G
4,4'-Methylenedi-o-toluidine ≥97.0% (by titrimetric analysis)
4,4'-Methylenedi-o-toluidine
Formel: C₁₅H₁₈N₂
MDL nummer: MFCD00126963
CAS-nummer: 838-88-0
EINECS: 212-658-8
FN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8d347f1a9e6cb2411fdedb8542fc3b6b14ff4e6&vid=e8d347f1a9e6cb2411fdedb8542fc3b6b14ff4e6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 94.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 152.0 to 159.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR