4,4'-Diisocyanato-3,3'-dimethylbiphenyl ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 3,3'-Dimethylbiphenyl-4,4'-diyl Diisocyanate

D2051-25G D2051-250G
TCIAD2051-25GEA 395 NOK
TCIAD2051-25G TCIAD2051-250G
4,4'-Diisocyanato-3,3'-dimethylbiphenyl ≥98.0% (by GC)
4,4'-Diisocyanato-3,3'-dimethylbiphenyl
Formel: C₁₆H₁₂N₂O₂
MW: 264.28 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00019908
CAS-nummer: 91-97-4
EINECS: 202-112-7

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white pellet
Purity (GC) min. 98.0 %
Melting point 69.0 to 73.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR