Methyl-3-(dimethylamino)benzoate ≥99.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 3-(Dimethylamino)benzoic acid methyl ester

D2069-10G D2069-25G
TCIAD2069-10GEA 985 NOK
TCIAD2069-10G TCIAD2069-25G
Methyl-3-(dimethylamino)benzoate ≥99.0% (by titrimetric analysis)
Methyl-3-(dimethylamino)benzoate
Formel: C₁₀H₁₃NO₂
Kokepunkt: 80°C
MDL nummer: MFCD00144731
CAS-nummer: 16518-64-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c015b36108642c2c2a9f12b8712e86c24776eb5&vid=0c015b36108642c2c2a9f12b8712e86c24776eb5&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 99.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR