Dioctadecyldimethylammonium bis(1,3-dithiole-2-thione-4,5-dithiolato)aurate(III)

Leverandør: TCI
Warning

Synonymer: Dimethyldioctadecylammonium Bis(1,3-dithiole-2-thione-4,5-dithiolato)aurate(III)

D2134-100MG
TCIAD2134-100MGEA 730 NOK
TCIAD2134-100MG
Dioctadecyldimethylammonium bis(1,3-dithiole-2-thione-4,5-dithiolato)aurate(III)
Dioctadecyldimethylammonium bis(1,3-dithiole-2-thione-4,5-dithiolato)aurate(III)
Formel: C₄₄H₈₀AuNS₁₀
MW: 1140,70 g/mol
MDL nummer: MFCD00144153
CAS-nummer: 120141-26-6

Bestill nå

Spesifikasjon Testresultater

Appearance Red to Dark blue to Black powder to crystal
Elemental analysis (Carbon) 44.00 to 48.50 %
Elemental analysis (Nitrogen) 1.00 to 1.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR