2-(4-Diethylamino-2-hydroxybenzoyl)benzoic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-Hydroxy-4-diethylamino-2'-hydroxycarbonylbenzophenone, 2-(4-(Diethylamino)-2-hydroxybenzoyl)benzoic acid

D2140-25G D2140-500G D2140-100G
TCIAD2140-25GEA 405 NOK
TCIAD2140-25G TCIAD2140-500G TCIAD2140-100G
2-(4-Diethylamino-2-hydroxybenzoyl)benzoic acid ≥98.0% (by GC, titration analysis)
2-(4-Diethylamino-2-hydroxybenzoyl)benzoic acid
Formel: C₁₈H₁₉NO₄
MW: 313.35 g/mol
Smeltepunkt: 180°C
MDL nummer: MFCD00134669
CAS-nummer: 5809-23-4
EINECS: 227-370-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR