cis-4,7,10,13,16,19-Docosahexaenoic acid ≥97.0% (by GC)

Leverandør: TCI

Synonymer: DHA, Docosa-4Z,7Z,10Z,13Z,16Z,19Z-Hexaenoic Acid

D2226-100MG
TCIAD2226-100MGEA 1700 NOK
TCIAD2226-100MG
cis-4,7,10,13,16,19-Docosahexaenoic acid ≥97.0% (by GC)
cis-4,7,10,13,16,19-Docosahexaenoic acid
Formel: C₂₂H₃₂O₂
Flash punkt: 62°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00065722
CAS-nummer: 6217-54-5
Merck Indeks: 14,03398
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58652d38432d61d29f05216a506432945c03c848&vid=46dae74431a7b45f604095bace77f90b63c181ee&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR