cis-2,6-Dimethylpiperazine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

D2360-25G D2360-500G
TCIAD2360-25GEA 595 NOK
TCIAD2360-25G TCIAD2360-500G
cis-2,6-Dimethylpiperazine ≥98.0% (by GC, titration analysis)
cis-2,6-Dimethylpiperazine
Formel: C₆H₁₄N₂
Kokepunkt: 160°C
MDL nummer: MFCD07772435
CAS-nummer: 21655-48-1
EINECS: 203-588-9
FN: 1325
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=864c7a4ed071d344483a57b5a85a7c8236d8db55&vid=864c7a4ed071d344483a57b5a85a7c8236d8db55&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 112.0 to 115.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR