Methyl-2,2-dimethoxypropionate ≥96.0% (by GC)

Leverandør: TCI

Synonymer: Methyl 2,2-dimethoxypropanoate

D2382-25G
TCIAD2382-25GEA 2225 NOK
TCIAD2382-25G
Methyl-2,2-dimethoxypropionate ≥96.0% (by GC)
Methyl-2,2-dimethoxypropionate
Formel: C₆H₁₂O₄
MW: 148.1571 g/mol
Kokepunkt: 70°C
MDL nummer: MFCD00048066
CAS-nummer: 10076-48-9
EINECS: 233-211-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a584c89073a8804e00979711ca1da03600486078&vid=a584c89073a8804e00979711ca1da03600486078&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR