Diphenyl-N-cyanocarbonimidate 97,0 HPLC, N

Leverandør: TCI
Danger

Synonymer: Diphenyl N-cyanocarbonimidate, N-Cyanocarbonimidic Acid Diphenyl Ester

D2454-5G
TCIAD2454-5GEA 1375 NOK
TCIAD2454-5G
Diphenyl-N-cyanocarbonimidate 97,0 HPLC, N
Diphenyl-N-cyanocarbonimidate
Formel: C₁₄H₁₀N₂O₂
MW: 238.25 g/mol
MDL nummer: MFCD00010380
CAS-nummer: 79463-77-7
EINECS: 427-300-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 158.0 to 162.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR