Dibenzyl diisopropylphosphoramidite ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

D2624-5G
TCIAD2624-5GEA 2650 NOK
TCIAD2624-5G
Dibenzyl diisopropylphosphoramidite ≥98.0% (by titrimetric analysis)
Dibenzyl diisopropylphosphoramidite
Formel: C₂₀H₂₈NO₂P
Flash punkt: 70°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00191988
CAS-nummer: 108549-23-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c576de1c944c9ad1e1ffbf2895e6bb4a31796149&vid=c576de1c944c9ad1e1ffbf2895e6bb4a31796149&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR