2,4-Dihydroxypyridine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Pyridine-2,4-diol, 2,4-Pyridindiol, 3-Deazauracil, 4-Hydroxy-2-pyridone

D2682-1G
TCIAD2682-1GEA 540 NOK
TCIAD2682-1G
2,4-Dihydroxypyridine ≥98.0% (by GC, titration analysis)
2,4-Dihydroxypyridine
Formel: C₅H₅NO₂
MW: 111,10 g/mol
Kokepunkt: 315 °C (1013 hPa)
Smeltepunkt: 275…277 °C
MDL nummer: MFCD00006273
CAS-nummer: 626-03-9
EINECS: 210-924-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92a7ddea7ceb3481d4e8d60babbe83b2d1536d80&vid=9cf6642a4124bda618567d07aba135339d05c124&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR