3,3-Diphenyl-1-propanol ≥98.0% (by GC)

Leverandør: TCI

D2760-25G D2760-5G
TCIAD2760-25GEA 8700 NOK
TCIAD2760-25G TCIAD2760-5G
3,3-Diphenyl-1-propanol ≥98.0% (by GC)
3,3-Diphenyl-1-propanol
Formel: C₁₅H₁₆O
Kokepunkt: 208°C
MDL nummer: MFCD00002930
CAS-nummer: 20017-67-8
EINECS: 243-466-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR