Diphenylphosphine oxide ≥98.0% (by GC)

Leverandør: TCI
Warning

D2826-25G D2826-5G
TCIAD2826-25GEA 1850 NOK
TCIAD2826-25G TCIAD2826-5G
Diphenylphosphine oxide ≥98.0% (by GC)
Diphenylphosphine oxide
Formel: C₁₂H₁₁OP
Kokepunkt: 105°C
MDL nummer: MFCD00002079
CAS-nummer: 4559-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3187317474ec552b6f7ea058e681b21185f35917&vid=3187317474ec552b6f7ea058e681b21185f35917&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR