1,1'-Dinaphthylamine ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Warning

Synonymer: 1,1'-Iminodinaphthalene

D2986-1G
TCIAD2986-1GEA 3550 NOK
TCIAD2986-1G
1,1'-Dinaphthylamine ≥98.0% (by HPLC, total nitrogen)
1,1'-Dinaphthylamine
Formel: C₂₀H₁₅N
MW: 269.35 g/mol
Kokepunkt: 257°C
CAS-nummer: 737-89-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 110.0 to 115.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR