(5S)-5,6-Dihydro-6,6-dimethyl-5-phenyl-2H-1,4-oxazin-2-one ≥95.0% (by GC)

Leverandør: TCI

D3259-1G
TCIAD3259-1GEA 7700 NOK
TCIAD3259-1G
(5S)-5,6-Dihydro-6,6-dimethyl-5-phenyl-2H-1,4-oxazin-2-one ≥95.0% (by GC)
(5S)-5,6-Dihydro-6,6-dimethyl-5-phenyl-2H-1,4-oxazin-2-one
Formel: C₁₂H₁₃NO₂
CAS-nummer: 723262-95-1

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 84.0 to 88.0 deg-C
Specific rotation [a]20/D +71.0 to +76.0 deg (C=2.7, Chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR