O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate ≥98.0% (by HPLC)

Leverandør: TCI
Danger

Synonymer: 1,1,3,3-Tetramethyl-2-(4-oxobenzo[d][1,2,3]triazin-3(4H)-yl)isouronium tetrafluoroborate, N,N,N',N'-Tetramethyl-O-(3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-yl)uronium Tetrafluoroborate, TDBTU

D3263-5G D3263-25G
TCIAD3263-5GEA 475 NOK
TCIAD3263-5G TCIAD3263-25G
O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate ≥98.0% (by HPLC)
O-(3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate
Formel: C₁₂H₁₆BF₄N₅O₂
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00077730
CAS-nummer: 125700-69-8
FN: 1759
ADR: 8,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR