1,1'-(Decane-1,10-diyl)bis[4-aza-1-azoniabicyclo[2.2.2]octane]dibromide ≥95.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 1,10-Bis[4-aza-1-azoniabicyclo[2.2.2]octan-1-yl]decane Dibromide

D3269-1G
TCIAD3269-1GEA 2275 NOK
TCIAD3269-1G
1,1'-(Decane-1,10-diyl)bis[4-aza-1-azoniabicyclo[2.2.2]octane]dibromide ≥95.0% (by titrimetric analysis)
1,1'-(Decane-1,10-diyl)bis[4-aza-1-azoniabicyclo[2.2.2]octane]dibromide
Formel: C₂₂H₄₄Br₂N₄
CAS-nummer: 94630-53-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR