Dimetylamin ∼11% ∼2.0 mol/L in methanol

Leverandør: TCI
Danger

D3292-500ML
TCIAD3292-500MLEA 700 NOK
TCIAD3292-500ML
Dimetylamin ∼11% ∼2.0 mol/L in methanol
Dimetylamin
Formel: C₂H₇N
Flash punkt: 5°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008288
CAS-nummer: 124-40-3
EINECS: 204-697-4
FN: 2924
ADR: 3,II
Merck Indeks: 14,03228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81428656b437a47821676b1ee64bdf4753dbdfb9&vid=621616780f834684ced12e4826a84e1f08a8d291&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Concentration (Neutralization titration) 11.0 to 13.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR