9,9-Dimethylfluorene-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI

D3332-1G
TCIAD3332-1GEA 2700 NOK
TCIAD3332-1G
9,9-Dimethylfluorene-2-carboxylic acid ≥98.0% (by GC, titration analysis)
9,9-Dimethylfluorene-2-carboxylic acid
Formel: C₁₆H₁₄O₂
CAS-nummer: 28320-62-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 194.0 to 198.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR