2'-Deoxycytidine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

D3583-1G D3583-5G
TCIAD3583-1GEA 350 NOK
TCIAD3583-1G TCIAD3583-5G
2'-Deoxycytidine ≥98.0% (by HPLC, titration analysis)
2'-Deoxycytidine
Formel: C₉H₁₃N₃O₄
MDL nummer: MFCD00006547
CAS-nummer: 951-77-9
EINECS: 213-454-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ee4cd46214109310caa1db6e79dfe60b5cd138b&vid=1ee4cd46214109310caa1db6e79dfe60b5cd138b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +56.0 to +61.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR