2'-Deoxy-5-methylcytidine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

D3610-100MG D3610-5G D3610-500MG
TCIAD3610-100MGEA 515 NOK
TCIAD3610-100MG TCIAD3610-5G TCIAD3610-500MG
2'-Deoxy-5-methylcytidine ≥98.0% (by HPLC, titration analysis)
2'-Deoxy-5-methylcytidine
Formel: C₁₀H₁₅N₃O₄
Smeltepunkt: 193°C
MDL nummer: MFCD00006549
CAS-nummer: 838-07-3
EINECS: 212-655-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +42.0 to +46.0 deg (C=1.4, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR