2'-Fluoro-2'-deoxycytidine ≥98.0% (by titrimetric analysis)

Leverandør: TCI

D3614-1G
TCIAD3614-1GEA 620 NOK
TCIAD3614-1G
2'-Fluoro-2'-deoxycytidine ≥98.0% (by titrimetric analysis)
2'-Fluoro-2'-deoxycytidine
Formel: C₉H₁₂FN₃O₄
Smeltepunkt: 167°C
MDL nummer: MFCD00057445
CAS-nummer: 10212-20-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +77.0 to +83.0 deg (C=1, Methanol) (calcd.on anh.substance)
Water max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR