2'-Deoxy-5-fluorocytidine ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

D3642-1G D3642-5G
TCIAD3642-1GEA 2150 NOK
TCIAD3642-1G TCIAD3642-5G
2'-Deoxy-5-fluorocytidine ≥98.0% (by HPLC, titration analysis)
2'-Deoxy-5-fluorocytidine
Formel: C₉H₁₂FN₃O₄
Smeltepunkt: 196°C
MDL nummer: MFCD00077348
CAS-nummer: 10356-76-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +65.0 to +75.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR