Dimethyl-cis-4-cyclohexene-1,2-dicarboxylate ≥97.0% (by GC)

Leverandør: TCI

D3703-5G
TCIAD3703-5GEA 610 NOK
TCIAD3703-5G
Dimethyl-cis-4-cyclohexene-1,2-dicarboxylate ≥97.0% (by GC)
Dimethyl-cis-4-cyclohexene-1,2-dicarboxylate
Formel: C₁₀H₁₄O₄
Kokepunkt: 142°C
MDL nummer: MFCD00209581
CAS-nummer: 4841-84-3

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR