Diphenyl (N-methoxy-N-methylcarbamoylmethyl)phosphonate ≥95.0% (by GC)

Leverandør: TCI

Synonymer: (N-Methoxy-N-methylcarbamoylmethyl)phosphonic Acid Diphenyl Ester

D3709-1G D3709-5G
TCIAD3709-1GEA 1500 NOK
TCIAD3709-1G TCIAD3709-5G
Diphenyl (N-methoxy-N-methylcarbamoylmethyl)phosphonate ≥95.0% (by GC)
Diphenyl (N-methoxy-N-methylcarbamoylmethyl)phosphonate
Formel: C₁₆H₁₈NO₅P
CAS-nummer: 367508-01-8

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 95.0 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR