Diethyl-cis-1,2-cyclohexanedicarboxylate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: cis-1,2-Cyclohexanedicarboxylic Acid Diethyl Ester

D3713-5G
TCIAD3713-5GEA 4450 NOK
TCIAD3713-5G
Diethyl-cis-1,2-cyclohexanedicarboxylate ≥98.0% (by GC)
Diethyl-cis-1,2-cyclohexanedicarboxylate
Formel: C₁₂H₂₀O₄
Kokepunkt: 119°C
MDL nummer: MFCD03938293
CAS-nummer: 17351-07-4

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR