Diethyl-cis-4-cyclohexene-1,2-dicarboxylate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: cis-4-Cyclohexene-1,2-dicarboxylic Acid Diethyl Ester

D3714-5G
TCIAD3714-5GEA 1450 NOK
TCIAD3714-5G
Diethyl-cis-4-cyclohexene-1,2-dicarboxylate ≥98.0% (by GC)
Diethyl-cis-4-cyclohexene-1,2-dicarboxylate
Formel: C₁₂H₁₈O₄
Kokepunkt: 114°C
CAS-nummer: 4841-85-4

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR