2,9-Diphenyl-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

D3849-1G
TCIAD3849-1GEA 3475 NOK
TCIAD3849-1G
2,9-Diphenyl-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)
2,9-Diphenyl-1,10-phenanthroline
Formel: C₂₄H₁₆N₂
MDL nummer: MFCD00044208
CAS-nummer: 25677-69-4

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 186.0 to 190.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR