1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt, DMPG Sodium Salt, Sodium (R)-2,3-bis(tetradecanoyloxy)propyl (2,3-dihydroxypropyl) phosphate, Dimyristoylphosphatidylglycerol Sodium Salt, Sodium (R)-2,3-Bis(tetradecanoyloxy)propyl 2,3-Dihydroxypropyl Phosphate

D3946-1G
TCIAD3946-1GEA 2850 NOK
TCIAD3946-1G
1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt ≥98.0% (by titrimetric analysis)
1,2-Dimyristoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt
Formel: C₃₄H₆₇O₁₀P
MW: 688.86 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00056649
CAS-nummer: 200880-40-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR