5,5''-Dibromo-2,2':5',2''-terthiophene ≥97.0% (by GC)

Leverandør: TCI

Synonymer: 2,5-Bis(5-bromo-2-thienyl)thiophene

D4050-1G D4050-5G
TCIAD4050-1GEA 1825 NOK
TCIAD4050-1G TCIAD4050-5G
5,5''-Dibromo-2,2':5',2''-terthiophene ≥97.0% (by GC)
5,5''-Dibromo-2,2':5',2''-terthiophene
Formel: C₁₂H₆Br₂S₃
MDL nummer: MFCD00219109
CAS-nummer: 98057-08-0

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 97.0 %
Melting point 156.0 to 160.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR