2,2-Dimethylvaleronitrile ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 2,2-Dimethylpentanenitrile

D4171-5G
TCIAD4171-5GEA 2450 NOK
TCIAD4171-5G
2,2-Dimethylvaleronitrile ≥98.0% (by GC)
2,2-Dimethylvaleronitrile
Formel: C₇H₁₃N
MW: 111.19 g/mol
Kokepunkt: 150°C
CAS-nummer: 20654-47-1
FN: 1993
ADR: 3 III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR