2,5-Dibromo-N-n-octyl-3,4-thiophenedicarboximide ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 1,3-Dibromo-5-octyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione, 1,3-Dibromo-5-n-octyl-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione

D4219-200MG
TCIAD4219-200MGEA 620 NOK
TCIAD4219-200MG
2,5-Dibromo-N-n-octyl-3,4-thiophenedicarboximide ≥98.0% (by GC)
2,5-Dibromo-N-n-octyl-3,4-thiophenedicarboximide
Formel: C₁₄H₁₇Br₂NO₂S
MW: 423,16 g/mol
MDL nummer: MFCD18804055
CAS-nummer: 566939-58-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR