1,3-Dichloroisoquinoline ≥98.0% (by GC)

Leverandør: TCI
Warning

D4285-1G
TCIAD4285-1GEA 890 NOK
TCIAD4285-1G
1,3-Dichloroisoquinoline ≥98.0% (by GC)
1,3-Dichloroisoquinoline
Formel: C₉H₅Cl₂N
MW: 198,05 g/mol
Kokepunkt: 307°C
MDL nummer: MFCD00034750
CAS-nummer: 7742-73-6

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR