Dimethyl [2-oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: [2-Oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonic Acid Dimethyl Ester

D4397-1G
TCIAD4397-1GEA 3425 NOK
TCIAD4397-1G
Dimethyl [2-oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonate ≥98.0% (by GC)
Dimethyl [2-oxo-3-[3-(trifluoromethyl)phenoxy]propyl]phosphonate
Formel: C₁₂H₁₄F₃O₅P
MW: 326,21 g/mol
MDL nummer: MFCD09054713
CAS-nummer: 54094-19-8

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR