D 131 ≥98.0% (by HPLC)

Leverandør: TCI
Warning

Synonymer: 2-Cyano-3-[4-[4-(2,2-diphenylvinyl)phenyl]-1,2,3,3a,4,8b-hexahydrocyclopenta[b]indol-7-yl]acrylic Acid, 2-Cyano-3-(4-(4-(2,2-diphenylvinyl)phenyl)-1,2,3,3a,4,8b-hexahydrocyclopenta[b]indol-7-yl)acrylic acid

D4431-50MG
TCIAD4431-50MGEA 1450 NOK
TCIAD4431-50MG
D 131 ≥98.0% (by HPLC)
D 131
Formel: C₃₅H₂₈N₂O₂
MW: 508,62 g/mol
MDL nummer: MFCD28100828
CAS-nummer: 652145-29-4

Bestill nå

Spesifikasjon Testresultater

Appearance Light red to Yellow to Red powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 5.30 to 5.65 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR