Dipotassium-(R)-1,1'-binaphthyl-2,2'-disulphonate ≥98.0% (by HPLC)

Leverandør: TCI

Synonymer: (R)-1,1'-Binaphthyl-2,2'-disulphonic acid dipotassium salt, (R)-BINSA Dipotassium salt

D4445-100MG
TCIAD4445-100MGEA 4925 NOK
TCIAD4445-100MG
Dipotassium-(R)-1,1'-binaphthyl-2,2'-disulphonate ≥98.0% (by HPLC)
Dipotassium-(R)-1,1'-binaphthyl-2,2'-disulphonate
Formel: C₂₀H₁₄O₆S₂
MW: 490,63 g/mol
CAS-nummer: 1092934-19-4

Bestill nå

Spesifikasjon Testresultater

Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Ion exchange titration) min. 95.0 %
Specific rotation [a]20/D +60.0 to +65.0 deg (C=1, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR