Daunorubicin hydrochloride 98,0 N_ASSAY_METHOD

Supplier: TCI
Danger

Synonyms: Daunorubicin HCl, Hydroxydaunorubicin Hydrochloride, (8S,10S)-8-Acetyl-10-(((2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride

D4532-20MG D4532-100MG
TCIAD4532-20MGEA 555 NOK
TCIAD4532-20MG TCIAD4532-100MG
Daunorubicin hydrochloride 98,0 N_ASSAY_METHOD
Daunorubicin hydrochloride
Formula: C₂₇H₂₉NO₁₁·HCl
MW: 563.98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04974507
CAS Number: 23541-50-6
EINECS: 245-723-4
UN: 2811
ADR: 6.1 III
Merck Index: 14,02832

Order Now

Specification Test Results

Appearance Orange to Red powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 98.0 % (calcd.on dried substance)
Specific rotation [a]20/D +250 to +275 deg (C=0.15, MeOH)
Drying loss max. 7.5 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR