4,4'-Diiodo-2,2'-dimethylbiphenyl ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 4,4'-Diiodo-2,2'-dimethyl-1,1'-biphenyl

D4608-1G D4608-5G
TCIAD4608-1GEA 1125 NOK
TCIAD4608-1G TCIAD4608-5G
4,4'-Diiodo-2,2'-dimethylbiphenyl ≥98.0% (by GC)
4,4'-Diiodo-2,2'-dimethylbiphenyl
Formel: C₁₄H₁₂I₂
MW: 434.06 g/mol
MDL nummer: MFCD13185944
CAS-nummer: 69571-02-4

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 100.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR