3-[Diethoxy(methyl)silyl]propyl methacrylate ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Methacrylic Acid 3-[Diethoxy(methyl)silyl]propyl Ester

D4679-25ML
TCIAD4679-25MLEA 475 NOK
TCIAD4679-25ML
3-[Diethoxy(methyl)silyl]propyl methacrylate ≥98.0% (by GC)
3-[Diethoxy(methyl)silyl]propyl methacrylate
Formel: C₁₂H₂₄O₄Si
MDL nummer: MFCD00053879
CAS-nummer: 65100-04-1

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR