2-(Diphenylphosphino)biphenyl ≥98.0% (by GC)

Leverandør: TCI
Warning

D4688-1G
TCIAD4688-1GEA 340 NOK
TCIAD4688-1G
2-(Diphenylphosphino)biphenyl ≥98.0% (by GC)
2-(Diphenylphosphino)biphenyl
Formel: C₂₄H₁₉P
MDL nummer: MFCD11559063
CAS-nummer: 13885-09-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR