Dimethyl-4-chloropyridine-2,6-dicarboxylate ≥98.0% (by GC, total nitrogen)

Leverandør: TCI
Warning

Synonymer: Dimethyl 4-chloropyridine-2,6-dicarboxylate, Dimethyl 4-Chloro-2,6-pyridinedicarboxylate, 4-Chloro-2,6-pyridinedicarboxylic acid dimethyl ester

D4693-1G
TCIAD4693-1GEA 915 NOK
TCIAD4693-1G
Dimethyl-4-chloropyridine-2,6-dicarboxylate ≥98.0% (by GC, total nitrogen)
Dimethyl-4-chloropyridine-2,6-dicarboxylate
Formel: C₉H₈ClNO₄
MDL nummer: MFCD02612781
CAS-nummer: 5371-70-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 140.0 to 144.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR