Dimetylsulfoksid ≥99.0% (by GC) for spektrofotometri

Leverandør: TCI
Warning

D5293-100ML D5293-500ML
TCIAD5293-100MLEA 220 NOK
TCIAD5293-100ML TCIAD5293-500ML
Dimetylsulfoksid ≥99.0% (by GC) for spektrofotometri
Dimetylsulfoksid
Formel: (CH₃)₂SO
Kokepunkt: 189°C
Flash punkt: 95°C
MDL nummer: MFCD00002089
CAS-nummer: 67-68-5
EINECS: 200-664-3
Merck Indeks: 14,03259
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f204e52e4620b1c83da07d4c7f3222879602703&vid=5443d9dcd1f268c5214fe906f8d4ccb4f0353461&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %
Absorbance (270nm) max. 0.40
Absorbance (275nm) max. 0.25
Absorbance (280nm) max. 0.20


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR