1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: Sodium (R)-2,3-Bis(oleoyloxy)propyl (2,3-Dihydroxypropyl) Phosphate, Dioleoylphosphatidylglycerol sodium salt, DOPG sodium salt

D5318-250MG
TCIAD5318-250MGEA 1750 NOK
TCIAD5318-250MG
1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt ≥98.0% (by titrimetric analysis)
1,2-Dioleoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt
Formel: C₄₂H₇₈NaO₁₀P
MW: 797.04 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD01310528
CAS-nummer: 67254-28-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=379556ffd712486bc5bf8194d03788eefbdf5e9e&vid=379556ffd712486bc5bf8194d03788eefbdf5e9e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 94.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR