5,5'-Diiodo-2,2'-dimethylbiphenyl ≥97.0% (by GC)

Leverandør: TCI

D5430-1G D5430-5G
TCIAD5430-1GEA 1525 NOK
TCIAD5430-1G TCIAD5430-5G
5,5'-Diiodo-2,2'-dimethylbiphenyl ≥97.0% (by GC)
5,5'-Diiodo-2,2'-dimethylbiphenyl
Formel: C₁₄H₁₂I₂
MW: 434.06 g/mol
CAS-nummer: 1392203-19-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19c3e64e240de4118c16294cd582ed840c98fff6&vid=19c3e64e240de4118c16294cd582ed840c98fff6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 115.0 to 119.0 deg-C

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR