2,2-Difluoropropionic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 2,2-Difluoropropanoic acid

D5531-1G
TCIAD5531-1GEA 540 NOK
TCIAD5531-1G
2,2-Difluoropropionic acid ≥98.0% (by GC, titration analysis)
2,2-Difluoropropionic acid
Formel: C₃H₄F₂O₂
MW: 110,06 g/mol
MDL nummer: MFCD03093764
CAS-nummer: 373-96-6
FN: 3261
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR