10-[4-[4,6-Di(1-adamantyl)-1,3,5-triazin-2-yl]phenyl]-10H-spiro[acridine-9,9'-fluorene] ≥99.0% (by HPLC), purified by sublimation

Leverandør: TCI

Synonymer: FA-TA

D5747-200MG
TCIAD5747-200MGEA 2250 NOK
TCIAD5747-200MG
10-[4-[4,6-Di(1-adamantyl)-1,3,5-triazin-2-yl]phenyl]-10H-spiro[acridine-9,9'-fluorene] ≥99.0% (by HPLC), purified by sublimation
10-[4-[4,6-Di(1-adamantyl)-1,3,5-triazin-2-yl]phenyl]-10H-spiro[acridine-9,9'-fluorene]
Formel: C₅₄H₅₀N₄
MW: 755,02 g/mol
CAS-nummer: 2250187-16-5

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystaline
Purity (HPLC) min. 99.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR