N,2-Diphenyl[60]fulleropyrrolidine 98,0 HPLC_ASSAY_METHOD (contains 5% Hexane at maximum)

Leverandør: TCI
Warning

D5757-100MG
TCIAD5757-100MGEA 5275 NOK
TCIAD5757-100MG
N,2-Diphenyl[60]fulleropyrrolidine 98,0 HPLC_ASSAY_METHOD (contains 5% Hexane at maximum)
N,2-Diphenyl[60]fulleropyrrolidine
Formel: C₇₄H₁₃N
MW: 915.93 g/mol
CAS-nummer: 1373934-14-5

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Amber to Dark red powder to crystal
Purity (HPLC) min. 98.0 area%
Hexane max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR