1,1-Ethanediol diacetate ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 1,1-Diacetoxyethane, Ethylidene Diacetate

E0030-25G
TCIAE0030-25GEA 1575 NOK
TCIAE0030-25G
1,1-Ethanediol diacetate ≥98.0% (by GC)
1,1-Ethanediol diacetate
Formel: C₆H₁₀O₄
Kokepunkt: 167°C
MDL nummer: MFCD00014980
CAS-nummer: 542-10-9
EINECS: 208-800-3
Merck Indeks: 14,03811

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR