N-Ethyl-o-toluidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 2-(Ethylamino)toluene, N-Ethyl-2-methylaniline

E0177-25ML E0177-500ML
TCIAE0177-25MLEA 220 NOK
TCIAE0177-25ML TCIAE0177-500ML
N-Ethyl-o-toluidine ≥98.0% (by GC, titration analysis)
N-Ethyl-o-toluidine
Formel: C₉H₁₃N
Kokepunkt: 214°C
Smeltepunkt: –15 °C
Tetthet: 0,95 g/cm³ (20 °C)
Flash punkt: 98°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00026695
CAS-nummer: 94-68-8
EINECS: 202-354-3
FN: 2754
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5e53228c7d3b5c565cc6753f1efde2947e5215b3&vid=5e53228c7d3b5c565cc6753f1efde2947e5215b3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light orange to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR