N-Ethyl-p-toluidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

E0179-25ML E0179-100ML
TCIAE0179-25MLEA 915 NOK
TCIAE0179-25ML TCIAE0179-100ML
N-Ethyl-p-toluidine ≥98.0% (by GC, titration analysis)
N-Ethyl-p-toluidine
Formel: C₉H₁₃N
Kokepunkt: 222°C
Flash punkt: 89°C
MDL nummer: MFCD00035793
CAS-nummer: 622-57-1
EINECS: 210-742-9
FN: 2754
ADR: 6.1,II

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR